Learn More
Sino Biological Recombinant Human KIR2DL4 Protein
Supplier:  Sino Biological 13052HCCS100
Encompass_Preferred
A DNA sequence encoding the human KIR2DL4 (ADY38409.1)(Met1-His242) was expressed with six amino acids (LEVLFQ) at the C-terminus was expressed and purified.
Catalog No. 50-161-8195