Learn More
Sino Biological Recombinant Human Resistin Protein (hFc Tag)
Supplier:  Sino Biological 13055H02H100
Encompass_Preferred
A DNA sequence encoding the human RETN (NP_065148.1) (Ser17-Pro108) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8199