Learn More
Sino Biological Recombinant Human IL-22 Protein (His Tag)
Supplier:  Sino Biological 13059H08Y50
Encompass_Preferred
A DNA sequence encoding the human IL22 (NP_065386.1) (Ala34-Ile179) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8203