Learn More
Sino Biological Recombinant Human IL-23 P19/IL23A Protein (mFc Tag)
Supplier:  Sino Biological 13062H05H50
Encompass_Preferred
A DNA sequence encoding the human IL23A (NP_057668.1) (Met1-Pro189) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-8211