Learn More
Sino Biological Recombinant Human PON3 Protein (50 Ser/Asn, His Tag)
Supplier:  Sino Biological 13063H07B100
Encompass_Preferred
A DNA sequence encoding the human PON3 (Q15166-1) (Met 1-Leu 354) (50 Ser/Asn) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-8212