Learn More
Sino Biological Recombinant Human GOLPH2/GOLM1 Protein (His Tag)
Supplier:  Sino Biological 13066H08H50
Encompass_Preferred
A DNA sequence encoding the human GOLM1 isoform 1 (Q8NBJ4-1) (Val 40-Leu 401) was fused with a polyhistidine tag at the C-terminus and a Met at the N-terminus.
Catalog No. 50-161-8217