Learn More
Sino Biological Recombinant Human TSPAN1 Protein (mFc Tag)
Supplier:  Sino Biological 13073H04H100
Encompass_Preferred
A DNA sequence encoding the human TSPAN1 (NP_005718.2) (Tyr110-Asn211) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-161-8222