Learn More
Sino Biological Recombinant Human TSPAN1 Protein (aa 110-211, rFc Tag)
Supplier:  Sino Biological 13073H15H100
Encompass_Preferred
A DNA sequence encoding the human TSPAN1 (O60635)(Tyr110-Asn211) was expressed with the Fc region of rabbit IgG at the N-terminus.
Catalog No. 50-161-8226