Learn More
Sino Biological Recombinant Human MTSS1 Protein (aa1-250, His & MBP Tag)
Supplier:  Sino Biological 13085H10E50
Encompass_Preferred
A DNA sequence encoding the human MTSS1 (EAW92073.1) N-terminal fragment (Met 1-Ser 250) was fused with an N-terminal polyhistidine-tagged MBP tag at the N-terminus.
Catalog No. 50-161-8238