Learn More
Sino Biological Recombinant Human IL-19 Protein (His Tag)
Supplier:  Sino Biological 13089H08H50
Encompass_Preferred
A DNA sequence encoding the human IL19 (AAF06663.1) (Met1-Ala177) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8242