Learn More
Sino Biological Recombinant Human Thioredoxin reductase 1/TRXR1 Protein (aa 161-647, His Tag), HPLC-verified
Supplier:  Sino Biological 13093H07E100
Encompass_Preferred
A DNA sequence encoding the human TXNRD1 isoform 1 (Q16881-1) (Tyr 161-Cys 647) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8251