Learn More
Sino Biological Recombinant Human IGSF11 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 13094H08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human IGSF11 (AAH34411.1) (Met 1-Gly 240) was fused with a polyhistidine tag at the C-terminus
Catalog No. 50-161-8254