Learn More
Sino Biological Recombinant Human PTP4A2 Protein (GST Tag)
Supplier:  Sino Biological 13097H09E100
Encompass_Preferred
A DNA sequence encoding the human PTP4A2 isoform 1 (Q12974-1) (Asn 2-Gln 167) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-8259