Learn More
Sino Biological Recombinant Human TLE3 Protein (aa 484-772, GST Tag)
Supplier:  Sino Biological 13099H09E100
Encompass_Preferred
A DNA sequence encoding the N-terminal fragment of human TLE3 (Q04726-1) (Ser 484-Tyr 772) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-8263