Learn More
Sino Biological Recombinant Human IGFBP7 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 13100H02H50
Encompass_Preferred
A DNA sequence encoding the human IGFBP7 (Q16270) (Met 1-Leu 282) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8267