Learn More
Sino Biological Recombinant Human IGFBP7 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 13100H08H50
Encompass_Preferred
A DNA sequence encoding the human IGFBP7 (Q16270) (Met 1-Leu 282) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8271