Learn More
Sino Biological Recombinant Human CGB7 Protein (His Tag)
Supplier:  Sino Biological 13120H08B100
Encompass_Preferred
A DNA sequence encoding the human CGB7 isoform 1 (P01233-1) (Met 1-Gln 165) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8281