Learn More
Sino Biological Recombinant Human WWP2 Protein (His & GST Tag)
Supplier:  Sino Biological 13125H20B100
Encompass_Preferred
A DNA sequence encoding the human WWP2 (O00308) (Met1-Glu870) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-8287