Learn More
Sino Biological Recombinant Human Mesothelin Protein (aa 296-580, hFc Tag), HPLC-verified
Supplier:  Sino Biological 13128H01H100
Encompass_Preferred
A DNA sequence encoding the human MSLN (Q13421-2) (Glu296-Gly580) was expressed,with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-8293