Learn More
Sino Biological Recombinant Human Mesothelin Protein (hFc Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 13128H01HB20
Encompass_Preferred
A DNA sequence encoding the human MSLN (Q13421-2) (Glu296-Gly580) was expressed with the fused Fc region of human IgG1 at the N-terminus. The expressed protein was biotinylated in vitro.
Catalog No. 50-161-8295