Learn More
Sino Biological Recombinant Human Mesothelin Protein (hFc Tag)
Supplier:  Sino Biological 13128H02H100
Encompass_Preferred
A DNA sequence encoding the human MSLN (Q13421-2) (Met1-Arg286) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8297