Learn More
Sino Biological Recombinant Human Mesothelin Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 13128H08H100
Encompass_Preferred
A DNA sequence encoding the human MSLN (Q13421-2) (Met1-Arg286) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8299