Learn More
Sino Biological Recombinant Human KIR2DL1 Protein (hFc Tag)
Supplier:  Sino Biological 13145H02H50
Encompass_Preferred
A DNA sequence encoding the human KIR2DL1 (NP_055033.2) extracellular domain (Met 1-His 245) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8321