Learn More
Sino Biological Recombinant Human CaMKI gamma/CAMK1G Protein (His & GST Tag)
Supplier:  Sino Biological 13148H20B50
Encompass_Preferred
A DNA sequence encoding the human CAMK1G isoform 1 (Q96NX5-1) (Met 1-Met 476) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-8328