Learn More
Sino Biological Recombinant Human p53R2 Protein (His Tag)
Supplier:  Sino Biological 13150H07E100
Encompass_Preferred
A DNA sequence encoding the human RRM2B (Q7LG56-1) (Met 1-Phe 351) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8330