Learn More
Sino Biological Recombinant Human CCDC134 Protein (His Tag)
Supplier:  Sino Biological 13168H08H100
Encompass_Preferred
A DNA sequence encoding the human CCDC134 (Q9H6E4) (Met 1-Leu 229) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8342