Learn More
Sino Biological Recombinant Human HEXB Protein (His Tag)
Supplier:  Sino Biological 13169H08H100
Encompass_Preferred
A DNA sequence encoding the human HEXB (P07686) (Ala43-Met556) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-8344