Learn More
Sino Biological Recombinant Human PTP alpha/PTPRA Protein (aa 174-793, His & GST Tag)
Supplier:  Sino Biological 13175H20B50
Encompass_Preferred
A DNA sequence encoding the human PTPRA isoform 2 (P18433-2) cytoplasmic domain (Ala 174-Lys 793) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-8355