Learn More
Sino Biological Recombinant Human OTUB2 Protein (His Tag)
Supplier:  Sino Biological 13177H07E100
Encompass_Preferred
A DNA sequence encoding the human OTUB2 (Q96DC9-1) (Met 1-His 234) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8358