Learn More
Sino Biological Recombinant Human Ki67/MKI67 Protein (GST Tag)
Supplier:  Sino Biological 13180H09E100
Encompass_Preferred
A DNA sequence encoding the mature form of human MKI67 (P46013-2) (Met1-Pro120) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-8360