Learn More
Sino Biological Recombinant Human NGFR/p75NTR Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 13184H08H100
Encompass_Preferred
A DNA sequence encoding the human NGFR (P08138) extracellular domain (Met 1-Asn 250) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8364