Learn More
Sino Biological Recombinant Human PHOSPHO1 Protein (His Tag)
Supplier:  Sino Biological 13187H07E100
Encompass_Preferred
A DNA sequence encoding the human PHOSPHO1 isoform 1 (Q8TCT1-1) (Met 1-Cys 267) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8370