Learn More
Sino Biological Recombinant Human STAT6 Protein (His Tag)
Supplier:  Sino Biological 13190H08B100
Encompass_Preferred
A DNA sequence encoding the human STAT6 isoform 1 (P42226-1) (Met 1-Trp 847) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8376