Learn More
Sino Biological Recombinant Human Parathyroid Hormone/PTH Protein (aa 32-65, GST Tag)
Supplier:  Sino Biological 13192H09E100
Encompass_Preferred
A DNA sequence encoding the human PTH (P01270) (Ser 32-Phe 65) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-8380