Learn More
Sino Biological Recombinant Human CD79A Protein (ECD, His Tag)
Supplier:  Sino Biological 13208H08H100
Encompass_Preferred
A DNA sequence encoding the human CD79A (NP_001774.1) (Met1-Arg143) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8403