Learn More
Eurofins Discoverx cAMP Hunter 1321N1 CALCR-RAMP3 Gs Cell Line, 2 vials. (95-0166C6)
Catalog No. 502003932
Encompass
The cAMP Hunter CALCR-RAMP3 Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. calcitonin) based activation of CALCR GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type CALCR, a naturally Gs coupled GPCR, alongside modulator protein RAMP3. Upon activation of CALCR-RAMP3, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. This product expresses CALCR alongside the receptor modifying protein RAMP3, which increases activity of the CALCR receptor For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3932 Supplier Eurofins Discoverx Supplier No. 950166C6
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter CALCR-RAMP3 Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. calcitonin) based activation of CALCR GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type CALCR, a naturally Gs coupled GPCR, alongside modulator protein RAMP3. Upon activation of CALCR-RAMP3, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. This product expresses CALCR alongside the receptor modifying protein RAMP3, which increases activity of the CALCR receptor For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.