Learn More
Eurofins Discoverx 1321N1 P2RY1 Gq Cell Line, 2 vials. (95-1018C6)
Catalog No. 502004113
Encompass
The 1321N1 P2RY1 Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. ATP) based activation of P2RY1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses P2RY1, a naturally Gq coupled GPCR. Upon activation of P2RY1, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-4113 Supplier Eurofins Discoverx Supplier No. 951018C6
Add to Cart
Add to Cart
Request a Quote
The 1321N1 P2RY1 Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. ATP) based activation of P2RY1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses P2RY1, a naturally Gq coupled GPCR. Upon activation of P2RY1, the resulting calcium activity can be fluorescently assayed using the Calcium No WashPLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.