Learn More
Sino Biological Recombinant Human Interferon alpha/beta receptor 1 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 13222H02H100
Encompass_Preferred
A DNA sequence encoding the human IFNAR1 isoform 1 (P17181-1) extracellular domain (Met 1-Lys 436) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8413