Learn More
Sino Biological Recombinant Human PACAP receptor/ADCYAP1R1 Protein (hFc Tag)
Supplier:  Sino Biological 13223H02H50
Encompass_Preferred
A DNA sequence encoding the human ADCYAP1R1 (P41586) extracellular domain (Met 1-Ala 155) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8418