Learn More
Sino Biological Recombinant Human p63 Protein (His & GST Tag)
Supplier:  Sino Biological 13226H20B100
Encompass_Preferred
A DNA sequence encoding the human TP63 isoform 1 (Q9H3D4-1) (Met 1-Glu 680) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-8427