Learn More
Sino Biological Recombinant Human CYB5R1 Protein (His Tag)
Supplier:  Sino Biological 13227H07E100
Encompass_Preferred
A DNA sequence encoding the human CYB5R1 (Q9UHQ9) (Leu 29-Tyr 305) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-8429