Learn More
Sino Biological Recombinant Human LRP10 Protein (hFc Tag)
Supplier:  Sino Biological 13228H02H100
Encompass_Preferred
A DNA sequence encoding the human LRP10 (Q7Z4F1-1) extracellular domain (Met 1-Lys 440) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8431