Learn More
Sino Biological Recombinant Human FDPS Protein (His Tag)
Supplier:  Sino Biological 13229H07E50
Encompass_Preferred
A DNA sequence encoding the human FDPS isoform b (NP_001129294.1) (Met 1-Lys 353) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8436