Learn More
Sino Biological Recombinant Human Procalcitonin/CALCA Protein (mFc Tag)
Supplier:  Sino Biological 13235H05H100
Encompass_Preferred
A DNA sequence encoding the human CALCA (NP_001732.1) (Met1-Asn141) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-8440