Learn More
Sino Biological Recombinant Human CART Protein
Supplier:  Sino Biological 13240HCCH100
Encompass_Preferred
A DNA sequence encoding the human CARTPT (NP_004282.1) (Met1-Leu116) was expressed with five amino acids (DDDDK) at the C-terminus.
Catalog No. 50-161-8448