Learn More
Sino Biological Recombinant Human Zinc Alpha 2 Glycoprotein/AZGP1 Protein (His Tag)
Supplier:  Sino Biological 13242H08H100
Encompass_Preferred
A DNA sequence encoding the human AZGP1 (P25311) (Met1-Ser298) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-8450