Learn More
Sino Biological Recombinant Human CST9L Protein (hFc Tag)
Supplier:  Sino Biological 13243H02H100
Encompass_Preferred
A DNA sequence encoding the human CST9L (Q9H4G1) (Met 1-His 147) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8452