Learn More
Sino Biological Recombinant Human IZUMO4 Protein (hFc Tag)
Supplier:  Sino Biological 13249H02H100
Encompass_Preferred
A DNA sequence encoding the human C19orf36 (Q1ZYL8) (Met1-His214) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8458