Learn More
Sino Biological Recombinant Human FKBP7 Protein (hFc Tag)
Supplier:  Sino Biological 13250H02H100
Encompass_Preferred
A DNA sequence encoding the human FKBP7 (Q9Y680-2) (Met1-Gln218) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8460