Learn More
Sino Biological Recombinant Human ERP27 Protein (mFc Tag)
Supplier:  Sino Biological 13256H05H100
Encompass_Preferred
A DNA sequence encoding the human ERP27 (Q96DN0) (Glu26-Pro269) was fused with Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-8464